Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019443.2 | NP_062316.1 | 2 | 313 | ||
GenBank mRNA | AI844651.1 | - | 2 | 183 | ||
AK007840.1 | - | 2 | 220 | |||
AK131584.1 | - | 512 | ||||
BC024364.1 | - | 2 | 222 | 5031206 | ||
CF383819.1 | - | 1 | 97 | |||
CT010398.1 | - | 2 | 165 | |||
Y07708.1 | - | 2 | 332 |