Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081695.2 | NP_001075164.1 | 11 | 1110 | ||
NM_001284197.1 | NP_001271126.1 | 9 | 951 | |||
NM_001284198.1 | NP_001271127.1 | 3 | 747 | |||
NM_001284199.1 | NP_001271128.1 | 3 | 574 | |||
NM_001284200.1 | NP_001271129.1 | 2 | 408 | |||
NM_019448.4 | NP_062321.1 | 10 | 964 | |||
GenBank mRNA | AF220524.1 | - | 9 | 895 | ||
AJ404467.1 | - | 9 | 936 | |||
AK010434.1 | - | 10 | 965 | |||
BC083147.1 | - | 9 | 949 | 30447906 | ||
EF051621.1 | - | 3 | 747 | |||
EF051622.1 | - | 3 | 574 | |||
EF051623.1 | - | 2 | 408 | |||
EF051624.1 | - | 11 | 1110 |