Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019456.2 | NP_062329.2 | 12, 13 | 1561 | ||
GenBank mRNA | AF020313.1 | - | 9, 10 | 1166 | ||
AK089987.1 | - | 4, 5 | 435 | |||
BC003991.1 | - | 9, 10 | 1212 | |||
BC023110.1 | - | 12, 13 | 1450 | 3707478 |