Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242363.2 | NP_001229292.1 | 13 | 4759 | ||
NM_001242364.2 | NP_001229293.1 | 12 | 4710 | |||
NM_001242365.2 | NP_001229294.1 | 12 | 4116 | |||
NM_199009.3 | NP_950174.1 | 12 | 4668 | |||
GenBank mRNA | AK014591.1 | - | 1 | 1550 | ||
AK122552.1 | - | 11 | 4417 |
Gene Symbol: Gm5901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195727.1 | NP_001182656.1 | 2 | 366 |