Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034905.2 | NP_001030077.1 | 2 | 141 | ||
NM_001039563.3 | NP_001034652.2 | 2 | 138 | |||
NM_001286493.1 | NP_001273422.1 | 2 | 141 | |||
GenBank mRNA | AK136627.1 | - | 2 | 142 | ||
AY552530.1 | - | 2 | 104 | |||
AY552531.1 | - | 2 | 107 | |||
BC145547.1 | - | 2 | 138 | 9054111 | ||
BC145549.1 | - | 2 | 135 | 9054113 | ||
BC145818.1 | - | 2 | 135 | 40131121 | ||
BC145820.1 | - | 2 | 135 | 40131123 | ||
DQ012055.1 | - | 2 | 99 |