Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019459.2 | NP_062332.2 | 22 | 3165 | ||
GenBank mRNA | AB513652.1 | - | 22 | 3037 | ||
AF168466.1 | - | 22 | 3037 | |||
AF172256.1 | - | 22 | 3165 | |||
AF191090.1 | - | 22 | 3493 | |||
AK141081.1 | - | 22 | 3188 |