Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025600.1 | NP_001020771.1 | 5 | 645 | ||
NM_018770.3 | NP_061240.3 | 5 | 645 | |||
NM_207675.2 | NP_997558.2 | 5 | 645 | |||
NM_207676.2 | NP_997559.1 | 5 | 645 | |||
GenBank mRNA | AB021964.1 | - | 2 | 174 | ||
AB021965.1 | - | 2 | 174 | |||
AB021966.1 | - | 2 | 174 | |||
AB021967.1 | - | 2 | 174 | |||
AB052293.1 | - | 5 | 645 | |||
AB064265.2 | - | 5 | 624 | |||
AB092414.1 | - | 5 | 624 | |||
AB183399.1 | - | 5 | 624 | |||
AB183400.1 | - | 5 | 624 | |||
AB183401.1 | - | 5 | 624 | |||
AB183402.1 | - | 5 | 624 | |||
AB377192.1 | - | 5 | 645 | |||
AF061260.1 | - | 4 | 482 | |||
AF434663.1 | - | 5 | 740 | |||
AF539424.1 | - | 5 | 637 | |||
AK013775.1 | - | 5 | 646 | |||
AK013911.1 | - | 5 | 739 | |||
AK053839.1 | - | 5 | 646 | |||
AY351388.1 | - | 5 | 624 | |||
BC057125.1 | - | 5 | 633 | 6414884 | ||
BC090970.1 | - | 5 | 824 | 6703838 | ||
BC094661.1 | - | 2 | 195 | |||
BC095986.1 | - | 5 | 637 | 5372277 | ||
DQ279856.1 | - | 5 | 624 | |||
JQ782219.1 | - | 5 | 624 |
Gene Symbol: Gm10677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048734.1 | - | 3 | 1835 |