Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_079835.2 | NP_524574.1 | 3 | 503 | ||
GenBank mRNA | AK136521.1 | - | 3 | 505 |