Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168568.1 | NP_001162040.1 | 8 | 1053 | ||
NM_001168569.1 | NP_001162041.1 | 8 | 1110 | |||
NM_001168571.1 | NP_001162043.1 | 8 | 1185 | |||
NM_018737.5 | NP_061207.1 | 8 | 1185 | |||
GenBank mRNA | AK032018.1 | - | 8 | 912 | ||
AK052854.1 | - | 8 | 1101 | |||
AK084595.1 | - | 8 | 982 | |||
AK085381.1 | - | 8 | 961 | |||
AK150966.1 | - | 8 | 1052 | |||
AK151597.1 | - | 8 | 1052 | |||
AK161564.1 | - | 8 | 1023 | |||
AK164206.1 | - | 8 | 1012 | |||
BC003257.1 | - | 8 | 893 | 3500778 | ||
U49385.1 | - | 8 | 1075 |