Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018854.4 | NP_061342.1 | 3 | 481 | ||
GenBank mRNA | AK002397.1 | - | 3 | 482 | ||
AK139942.1 | - | 3 | 518 | |||
AK213963.1 | - | 2 | 351 | |||
AY082613.1 | - | 2 | 201 | |||
BC002040.1 | - | 3 | 472 | 3590790 | ||
U38981.1 | - | 314 |