Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018868.2 | NP_061356.2 | 10 | 1213 | ||
GenBank mRNA | AF053232.1 | - | 10 | 1118 | ||
AK044216.1 | - | 1210 | ||||
AK133959.1 | - | 10 | 1211 | |||
AK134286.1 | - | 10 | 1214 | |||
BC076604.1 | - | 10 | 1214 | 30630088 | ||
BC085135.1 | - | 10 | 1183 | 5718488 |