Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285870.1 | NP_001272799.1 | 8 | 1897 | ||
NM_001285871.1 | NP_001272800.1 | 8 | 2082 | |||
NM_001285873.1 | NP_001272802.1 | 8 | 1897 | |||
NM_018880.3 | NP_061368.1 | 8 | 1871 | |||
GenBank mRNA | AB030912.1 | - | 8 | 1898 | ||
AF220019.1 | - | 8 | 1790 | |||
AK019165.1 | - | 8 | 1832 | |||
AK144674.1 | - | 8 | 1800 | |||
AK170029.1 | - | 8 | 1835 | |||
BC003347.1 | - | 8 | 1911 | |||
BC034263.1 | - | 8 | 1836 | 4503884 |