Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uso1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019490.1 | NP_062363.1 | 5 | 486 | ||
GenBank mRNA | AF096868.1 | - | 4 | 274 | ||
AK144638.1 | - | 5 | 477 | |||
AK156070.1 | - | 5 | 479 | |||
AK166587.1 | - | 6 | 446 | |||
AK172425.1 | - | 5 | 477 | |||
BC016069.1 | - | 5 | 486 | 4480992 |