Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019536.1 | NP_062409.1 | 66 | 11679 | ||
GenBank mRNA | AK147443.1 | - | 18 | 5304 |