Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019547.2 | NP_062420.2 | 2 | 472 | ||
GenBank mRNA | AK146026.1 | - | 2 | 446 | ||
AK162141.1 | - | 2 | 445 | |||
AK166609.1 | - | 2 | 473 | |||
AK172625.1 | - | 2 | 443 | |||
BC006687.1 | - | 1 | 981 | |||
BC085307.1 | - | 2 | 416 | 6396103 |