Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027204.2 | NP_081480.2 | 4 | 407 | ||
GenBank mRNA | AK002757.1 | - | 4 | 417 | ||
AK005328.1 | - | 4 | 438 | |||
AK028168.1 | - | 4 | 418 | |||
AK134816.1 | - | 4 | 440 | |||
AK166969.1 | - | 4 | 436 | |||
AK168337.1 | - | 4 | 410 | |||
BC005712.1 | - | 4 | 407 | 3582724 | ||
BC019565.1 | - | 4 | 405 | 4219731 |
Gene Symbol: Gm16755 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131899.1 | - | 2 | 165 |