Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019713.4 | NP_062687.1 | 1 | 213 | ||
GenBank mRNA | AF132851.1 | - | 1 | 46 | ||
BC002173.1 | - | 1 | 140 | 3486727 | ||
CJ156337.1 | - | 1 | 214 | |||
CX224234.1 | - | 1 | 165 |