Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291161.1 | NP_001278090.1 | 2 | 346 | ||
NM_019818.5 | NP_062792.2 | 2 | 427 | |||
GenBank mRNA | AF223950.1 | - | 2 | 393 | ||
AK011745.1 | - | 2 | 379 | |||
AK012130.2 | - | 2 | 381 | |||
AK017389.1 | - | 2 | 380 | |||
AK028132.1 | - | 2 | 379 | |||
AK153837.1 | - | 2 | 382 | |||
BC022610.1 | - | 2 | 363 | |||
BC055279.1 | - | 2 | 391 | 5688754 | ||
BF470824.1 | - | 2 | 403 | |||
CA751902.1 | - | 2 | 319 |