Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019875.2 | NP_063928.2 | 5 | 1180 | ||
GenBank mRNA | AB045382.2 | - | 4 | 1000 | ||
AB093298.1 | - | 5 | 1162 | |||
AF216495.1 | - | 5 | 1173 | |||
AK044140.1 | - | 5 | 1158 | |||
BC053014.1 | - | 5 | 1133 | 5686698 |