Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019721.2 | NP_062695.2 | 4 | 877 | ||
GenBank mRNA | AK085189.1 | - | 4 | 864 | ||
AK151616.1 | - | 4 | 864 | |||
AK153113.1 | - | 4 | 864 | |||
AK178994.1 | - | 2 | 115 | |||
BC012526.1 | - | 4 | 861 | 3981018 |