Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019757.1 | NP_062731.1 | 6 | 551 | ||
GenBank mRNA | AF083809.1 | - | 6 | 551 | ||
AK051993.1 | - | 6 | 634 | |||
AK081476.1 | - | 6 | 605 | |||
AK083686.1 | - | 614 | ||||
AK154822.1 | - | 6 | 635 | |||
AK155370.1 | - | 6 | 633 | |||
AK163951.1 | - | 6 | 610 | |||
AK172679.1 | - | 6 | 645 | |||
AK206176.1 | - | 1 | 24 | |||
BC006616.1 | - | 6 | 632 | 3967009 |