Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110004F10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019772.2 | NP_062746.1 | 5 | 593 | ||
GenBank mRNA | AB025407.1 | - | 5 | 593 | ||
AK003415.1 | - | 5 | 634 | |||
AK144059.1 | - | 5 | 620 | |||
AK167571.1 | - | 5 | 622 | |||
AK168206.1 | - | 5 | 627 | |||
AK169309.1 | - | 5 | 620 | |||
AK170172.1 | - | 3 | 351 | |||
BC072575.1 | - | 5 | 629 | 5715413 |