Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019694.1 | NP_062668.1 | 10 | 1728 | ||
GenBank mRNA | AB041582.1 | - | 10 | 1741 | ||
AF061026.1 | - | 10 | 1728 | |||
AK133849.1 | - | 10 | 1748 | |||
AK161638.1 | - | 4 | 1450 | |||
BC023862.1 | - | 10 | 1725 | |||
BC046326.1 | - | 10 | 1735 | |||
BC055865.1 | - | 10 | 1746 | |||
BC061115.1 | - | 10 | 1741 | 6533034 | ||
BC061690.1 | - | 10 | 1728 |