Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019694.1 | NP_062668.1 | 12 | 2025 | ||
GenBank mRNA | AB041582.1 | - | 12 | 2038 | ||
AF061026.1 | - | 12 | 2025 | |||
AK133849.1 | - | 12 | 2045 | |||
AK161638.1 | - | 5 | 2784 | |||
AK199956.1 | - | 1 | 47 | |||
AK207004.1 | - | 2 | 152 | |||
AK210262.1 | - | 1 | 79 | |||
AK215386.1 | - | 1 | 106 | |||
AK217209.1 | - | 1 | 77 | |||
BC023862.1 | - | 12 | 2022 | |||
BC046326.1 | - | 12 | 2032 | |||
BC055865.1 | - | 12 | 2043 | |||
BC061115.1 | - | 12 | 2038 | 6533034 | ||
BC061690.1 | - | 12 | 2024 |