Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019780.1 | NP_062754.1 | 2, 3 | 250 | ||
GenBank mRNA | AB578899.1 | - | 1, 2 | 22 | ||
AB578901.1 | - | 1, 2 | 22 | |||
AF193794.1 | - | 2, 3 | 250 | |||
AK004103.1 | - | 3, 4 | 267 | |||
AK008248.1 | - | 3, 4 | 270 | |||
AK036426.1 | - | 2, 3 | 238 | |||
AK150330.1 | - | 2, 3 | 260 | |||
BC005663.1 | - | 2, 3 | 239 | 2645432 |