Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019794.4 | NP_062768.1 | 3 | 372 | ||
GenBank mRNA | AB028853.1 | - | 3 | 373 | ||
AK031858.1 | - | 3 | 439 | |||
AK077672.1 | - | 3 | 443 | |||
AK083208.1 | - | 3 | 440 | |||
AK163255.1 | - | 3 | 371 | |||
AK169169.1 | - | 3 | 432 | |||
AK212721.1 | - | 1 | 83 | |||
BC003420.1 | - | 3 | 445 | 2582183 |