Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019786.4 | NP_062760.3 | 11 | 1449 | ||
GenBank mRNA | AF145705.1 | - | 10 | 1301 | ||
AF191839.1 | - | 10 | 1328 | |||
AK004649.2 | - | 11 | 1403 | |||
AK083214.1 | - | 11 | 1397 | |||
AK168193.1 | - | 11 | 1431 | |||
BC129910.1 | - | 10 | 1361 | 40111752 | ||
BC129911.1 | - | 10 | 1361 | 40111754 |