Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110159.1 | NP_001103629.1 | 3 | 787 | ||
NM_019761.6 | NP_062735.4 | 2 | 678 | |||
GenBank mRNA | AF156958.2 | - | 2 | 619 | ||
AK003254.1 | - | 2 | 654 | |||
AK168314.1 | - | 2 | 638 | |||
AW908959.1 | - | 1 | 288 | |||
BC023672.1 | - | 2 | 636 | 5039139 |