Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019656.3 | NP_062630.2 | 1 | 170 | ||
GenBank mRNA | AA458002.1 | - | 1 | 170 | ||
AF053454.1 | - | 1 | 117 | |||
AK146673.1 | - | 1 | 75 | |||
AK149204.1 | - | 1 | 80 | |||
AK160313.1 | - | 1 | 64 | |||
AK167384.1 | - | 1 | 66 | |||
BC003733.1 | - | 1 | 104 | 3590862 | ||
BC110314.1 | - | 1 | 79 | 5344878 |