Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020009.2 | NP_064393.2 | 5 | 669 | ||
GenBank mRNA | AF152838.1 | - | 5 | 644 | ||
AK036004.1 | - | 3 | 1855 | |||
AK143381.1 | - | 2 | 80 | |||
AK172584.1 | - | 5 | 660 | |||
BC021454.1 | - | 5 | 636 | |||
BC043920.1 | - | 5 | 626 | |||
BC112904.1 | - | 5 | 627 | 6465126 |