Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2l6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019949.2 | NP_064333.2 | 4 | 502 | ||
GenBank mRNA | AF159230.1 | - | 4 | 488 | ||
AK010942.1 | - | 4 | 481 | |||
AK013452.1 | - | 4 | 492 | |||
AK152010.1 | - | 4 | 482 | |||
AK152066.1 | - | 4 | 454 | |||
AK152113.1 | - | 4 | 503 | |||
AK168695.1 | - | 4 | 440 | |||
AK171629.1 | - | 4 | 451 | |||
BC008238.1 | - | 4 | 470 | 2651702 | ||
CT010270.1 | - | 4 | 421 |