Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020277.2 | NP_064673.2 | 4 | 488 | ||
GenBank mRNA | AB039952.1 | - | 3 | 405 | ||
AF228681.1 | - | 4 | 488 | |||
AJ271092.2 | - | 3 | 401 | |||
AY280364.1 | - | 4 | 452 | |||
AY280365.1 | - | 4 | 452 | |||
BC133712.1 | - | 3 | 405 | 40143371 |