Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021546.3 | NP_067521.2 | 3, 4 | 243 | ||
GenBank mRNA | AB039948.1 | - | 1, 2 | 64 | ||
AK013520.1 | - | 3, 4 | 242 | |||
AK044296.1 | - | 3, 4 | 235 | |||
AK158225.1 | - | 3, 4 | 245 | |||
BC126874.1 | - | 1, 2 | 57 | 40044848 | ||
BC126875.1 | - | 1, 2 | 57 | 40044849 |