Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021546.3 | NP_067521.2 | 8 | 689 | ||
GenBank mRNA | AB039948.1 | - | 6 | 510 | ||
AK013520.1 | - | 8 | 688 | |||
AK044296.1 | - | 8 | 681 | |||
AK158225.1 | - | 8 | 691 | |||
BC126874.1 | - | 6 | 503 | 40044848 | ||
BC126875.1 | - | 6 | 503 | 40044849 |