Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tob2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020507.3 | NP_065253.1 | 2 | 1426 | ||
GenBank mRNA | AK049766.1 | - | 2 | 1374 | ||
AK122537.1 | - | 1 | 2631 | |||
AK143542.1 | - | 2 | 1403 | |||
BC011163.1 | - | 1 | 1308 | 4194136 | ||
BC021869.1 | - | 2 | 1360 | |||
BC094031.1 | - | 2 | 1357 | 4945656 |