Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020626.2 | NP_065651.1 | 5 | 505 | ||
GenBank mRNA | AF178085.1 | - | 5 | 505 | ||
AK002337.1 | - | 5 | 451 | |||
AK148534.1 | - | 5 | 502 | |||
BC049912.1 | - | 5 | 427 | 4973401 |