Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020575.2 | NP_065600.1 | 7 | 1994 | ||
GenBank mRNA | AF155739.1 | - | 7 | 1994 | ||
AK034109.1 | - | 1984 | ||||
AK045853.1 | - | 2 | 83 | |||
AK143177.1 | - | 7 | 2016 | |||
AK166666.1 | - | 7 | 2001 | |||
AK167246.1 | - | 7 | 1976 | |||
AK168555.1 | - | 7 | 1997 | |||
BC025029.1 | - | 7 | 1958 | 4935124 |