Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem183a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042485.1 | NP_001035950.1 | 8 | 1115 | ||
NM_020588.2 | NP_065613.1 | 8 | 1118 | |||
GenBank mRNA | AB041592.1 | - | 8 | 1074 | ||
AK004922.2 | - | 8 | 1099 | |||
AK005019.1 | - | 8 | 1035 | |||
AK007779.1 | - | 8 | 1097 | |||
AK152852.1 | - | 8 | 1058 | |||
AK168331.1 | - | 8 | 1060 | |||
BC003210.1 | - | 8 | 1043 | 3156957 |