Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serinc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025371.2 | NP_001020542.1 | 6, 7 | 1040 | ||
GenBank mRNA | AK044479.1 | - | 6, 7 | 1068 | ||
BC083132.1 | - | 6, 7 | 1040 | 6466398 |