Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029929.2 | NP_001025100.1 | 10 | 2115 | ||
GenBank mRNA | AK029988.1 | - | 10 | 2116 | ||
AK128979.1 | - | 9 | 2422 | |||
AK171931.1 | - | 10 | 1964 | |||
BC038060.1 | - | 7 | 725 | |||
BC151086.1 | - | 10 | 1787 | 9087999 |