Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113527.1 | NP_001106999.1 | 1 | 225 | ||
NM_001291220.1 | NP_001278149.1 | 2 | 209 | |||
NM_001291221.1 | NP_001278150.1 | 2 | 209 | |||
NM_020583.5 | NP_065608.2 | 2 | 416 | |||
GenBank mRNA | AF217484.1 | - | 2 | 175 | ||
AK005528.1 | - | 2 | 208 | |||
AK008139.1 | - | 1 | 226 | |||
AK008171.1 | - | 1 | 226 | |||
AK008353.1 | - | 1 | 228 | |||
AK136756.1 | - | 2 | 342 | |||
BC022751.1 | - | 2 | 363 | 4912320 | ||
CT010213.1 | - | 1 | 113 | |||
W97502.1 | - | 2 | 380 |