Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sertad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038625.1 | NP_001033714.1 | 2 | 994 | ||
NM_021372.2 | NP_067347.2 | 2 | 1205 | |||
GenBank mRNA | AF366403.1 | - | 1 | 309 | ||
AK076787.1 | - | 2 | 898 | |||
AK129059.1 | - | 1 | 154 | |||
AK132846.1 | - | 2 | 974 | |||
AK132849.1 | - | 2 | 813 | |||
AK142018.1 | - | 2 | 974 | |||
AK146621.1 | - | 2 | 995 | |||
AK146897.1 | - | 2 | 996 | |||
AK147674.1 | - | 3 | 1381 | |||
AK195500.1 | - | 11 | ||||
BC008257.1 | - | 1 | 151 | |||
BC014726.1 | - | 2 | 1037 | 4240282 |