Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021419.1 | NP_067394.1 | 2 | 285 | ||
GenBank mRNA | AJ242721.1 | - | 229 | |||
AK076180.1 | - | 2 | 265 | |||
AK147168.1 | - | 2 | 307 | |||
AK219887.1 | - | 2 | 60 | |||
BC021778.1 | - | 2 | 285 | 4239442 | ||
CT010295.1 | - | 2 | 148 |