Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021473.3 | NP_067448.1 | 7 | 1143 | ||
GenBank mRNA | AF225564.1 | - | 7 | 1048 | ||
AK005162.1 | - | 4 | 584 | |||
AK005376.1 | - | 7 | 1078 | |||
AK009462.1 | - | 7 | 920 | |||
AK011157.1 | - | 4 | 584 | |||
AK011209.1 | - | 4 | 584 | |||
AK011221.1 | - | 4 | 583 | |||
AK011321.1 | - | 7 | 1072 | |||
AK011388.1 | - | 7 | 1098 | |||
AK011667.1 | - | 7 | 1099 | |||
AK011794.1 | - | 7 | 1072 | |||
AK011856.1 | - | 7 | 1099 | |||
AK011906.1 | - | 7 | 1077 | |||
AK011908.1 | - | 7 | 1112 | |||
AK011918.1 | - | 7 | 1098 | |||
AK128906.1 | - | 7 | 865 | |||
AK144198.1 | - | 7 | 1099 | |||
AK146081.1 | - | 7 | 1097 | |||
AK146278.1 | - | 7 | 1099 | |||
AK152086.1 | - | 7 | 1078 | |||
AK159573.1 | - | 7 | 1095 | |||
AK164461.1 | - | 7 | 1098 | |||
AK168950.1 | - | 7 | 1094 | |||
AK181349.1 | - | 3 | 318 | |||
AY207463.1 | - | 7 | 1120 | |||
BC003850.1 | - | 7 | 926 | |||
BC039926.1 | - | 7 | 1092 | 3156658 | ||
BC046762.1 | - | 7 | 1049 | 5693736 |