Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042491.2 | NP_001035956.1 | 13 | 1687 | ||
NM_001289517.1 | NP_001276446.1 | 13 | 1687 | |||
NM_001289518.1 | NP_001276447.1 | 13 | 1726 | |||
NM_001289519.1 | NP_001276448.1 | 13 | 1726 | |||
NM_001289520.1 | NP_001276449.1 | 13 | 1702 | |||
NM_021505.3 | NP_067480.2 | 13 | 1726 | |||
GenBank mRNA | AK012579.1 | - | 6 | 626 | ||
AK031432.1 | - | 13 | 1651 | |||
AK049719.1 | - | 13 | 1680 | |||
AK075969.1 | - | 13 | 1663 | |||
AK088327.1 | - | 13 | 1689 | |||
AK148679.1 | - | 13 | 1642 | |||
AK151314.1 | - | 13 | 1633 | |||
AK151905.1 | - | 13 | 1672 | |||
AK152528.1 | - | 13 | 1603 | |||
AK152981.1 | - | 13 | 1634 | |||
AK153373.1 | - | 13 | 1633 | |||
AK153422.1 | - | 13 | 1633 | |||
AK159204.1 | - | 13 | 1633 | |||
AK159713.1 | - | 13 | 1641 | |||
AK159916.1 | - | 13 | 1665 | |||
AK167561.1 | - | 12 | 2602 | |||
AK168135.1 | - | 13 | 1650 | |||
AK168737.1 | - | 13 | 1627 | |||
AK168814.1 | - | 13 | 1665 | |||
AK169196.1 | - | 13 | 1689 | |||
BC010339.1 | - | 13 | 1642 | 2648173 | ||
BC046566.1 | - | 13 | 1666 | 2650790 | ||
BC046804.1 | - | 13 | 1624 | 6404923 |