Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042491.2 | NP_001035956.1 | 14 | 1791 | ||
NM_001289517.1 | NP_001276446.1 | 14 | 1791 | |||
NM_001289518.1 | NP_001276447.1 | 14 | 1830 | |||
NM_001289519.1 | NP_001276448.1 | 14 | 1830 | |||
NM_001289520.1 | NP_001276449.1 | 14 | 1806 | |||
NM_021505.3 | NP_067480.2 | 14 | 1830 | |||
GenBank mRNA | AB041593.1 | - | 14 | 1774 | ||
AK012579.1 | - | 7 | 730 | |||
AK031432.1 | - | 14 | 1755 | |||
AK049719.1 | - | 14 | 1784 | |||
AK075969.1 | - | 14 | 1767 | |||
AK088327.1 | - | 14 | 1793 | |||
AK148679.1 | - | 14 | 1746 | |||
AK151314.1 | - | 14 | 1737 | |||
AK151905.1 | - | 14 | 1776 | |||
AK152981.1 | - | 14 | 1738 | |||
AK153373.1 | - | 14 | 1737 | |||
AK153422.1 | - | 14 | 1737 | |||
AK159204.1 | - | 14 | 1737 | |||
AK159713.1 | - | 14 | 1745 | |||
AK159916.1 | - | 14 | 1769 | |||
AK167561.1 | - | 13 | 2706 | |||
AK168135.1 | - | 14 | 1754 | |||
AK168737.1 | - | 14 | 1731 | |||
AK168814.1 | - | 14 | 1769 | |||
AK169196.1 | - | 14 | 1793 | |||
AK209720.1 | - | 2 | 111 | |||
BC010339.1 | - | 14 | 1746 | 2648173 | ||
BC046804.1 | - | 14 | 1728 | 6404923 |