Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021510.2 | NP_067485.1 | 5, 6 | 663 | ||
GenBank mRNA | AK088169.1 | - | 5, 6 | 616 | ||
AK161491.1 | - | 761 | ||||
AK178902.1 | - | 2, 3 | 270 | |||
BC042187.1 | - | 5, 6 | 598 | 5376052 | ||
BC056224.1 | - | 4, 5 | 593 | 6515270 | ||
BU058854.1 | - | 3, 4 | 416 | |||
Y14196.1 | - | 5, 6 | 621 |