Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021511.2 | NP_067486.2 | 1 | 564 | ||
GenBank mRNA | AB041589.1 | - | 1 | 530 | ||
AK010178.1 | - | 1 | 513 | |||
AK017683.1 | - | 1 | 516 | |||
AK019913.1 | - | 1 | 512 | |||
AK122207.2 | - | 1 | 525 | |||
AK142041.1 | - | 1 | 565 | |||
BC003481.1 | - | 1 | 540 | 2811004 | ||
BC055925.1 | - | 1 | 524 | 4235851 |