Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021518.3 | NP_067493.1 | 4 | 472 | ||
GenBank mRNA | AB232585.1 | - | 4 | 253 | ||
AK042862.1 | - | 4 | 442 | |||
AK079071.1 | - | 4 | 473 | |||
AK135421.1 | - | 4 | 456 | |||
AK146179.1 | - | 4 | 457 | |||
AK150030.1 | - | 4 | 504 | |||
AK159607.1 | - | 4 | 493 | |||
AK160333.1 | - | 4 | 473 | |||
AK163320.1 | - | 4 | 455 | |||
BC058382.1 | - | 4 | 470 | 6820798 | ||
X95403.1 | - | 4 | 460 |