Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021518.3 | NP_067493.1 | 6 | 642 | ||
GenBank mRNA | AB232585.1 | - | 6 | 423 | ||
AK042862.1 | - | 6 | 612 | |||
AK079071.1 | - | 6 | 643 | |||
AK135421.1 | - | 6 | 626 | |||
AK146179.1 | - | 6 | 627 | |||
AK150030.1 | - | 6 | 674 | |||
AK159607.1 | - | 6 | 663 | |||
AK160333.1 | - | 6 | 643 | |||
AK163320.1 | - | 6 | 625 | |||
AK208722.1 | - | 2 | 146 | |||
AK212917.1 | - | 2 | 168 | |||
BC058382.1 | - | 6 | 640 | 6820798 | ||
X95403.1 | - | 6 | 630 |